siRNA Details
| Gene Symbol: VPS13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018088.2 | NP_001018098.1 | 61 | 8185 | ||
| NM_017684.4 | NP_060154.3 | 59 | 8056 | |||
| NM_018080.3 | NP_060550.2 | 59 | 8056 | |||
| NM_020821.2 | NP_065872.1 | 61 | 8185 | |||
| XM_011521713.2 | XP_011520015.1 | 61 | 8146 | |||
| GenBank mRNA | AJ608770.1 | - | 59 | 8039 | ||
| AJ608771.1 | - | 61 | 8168 | |||
| AJ626860.1 | - | 59 | 8039 | |||
| AJ626861.1 | - | 61 | 8168 | |||
| AK001243.1 | - | 2 | 231 | |||
| AK128650.1 | - | 15 | 2599 | |||
| CR936716.1 | - | 10 | 1666 | |||

VPS13C
Genomic Map