siRNA Details
| Gene Symbol: Zfp521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145492.4 | NP_663467.1 | 8 | 4251 | ||
| GenBank mRNA | AK052655.1 | - | 8 | 4155 | ||
| AK077697.1 | - | 5 | 1073 | |||
| AK081096.1 | - | 6 | 1705 | |||
| AK131130.1 | - | 7 | 4336 | |||
| AY147406.1 | - | 8 | 4154 | |||
| BC021376.1 | - | 8 | 4067 | 5038671 | ||
| BC023458.1 | - | 5 | 787 | |||

Zfp521
Genomic Map