siRNA Details
| Gene Symbol: Zfp521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145492.4 | NP_663467.1 | 7, 8 | 4142 | ||
| GenBank mRNA | AK052655.1 | - | 7, 8 | 4046 | ||
| AK077697.1 | - | 4, 5 | 964 | |||
| AK081096.1 | - | 5, 6 | 1596 | |||
| AK131130.1 | - | 6, 7 | 4227 | |||
| AY147406.1 | - | 7, 8 | 4045 | |||
| BC021376.1 | - | 7, 8 | 3958 | 5038671 | ||
| BC023458.1 | - | 4, 5 | 678 | |||

Zfp521
Genomic Map