siRNA Details
| Gene Symbol: Olfr1303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146402.2 | NP_666514.2 | 1 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1303| Gene Symbol: Olfr1303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146402.2 | NP_666514.2 | 1 | 466 | ||