siRNA Details
| Gene Symbol: Olfr223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146429.1 | NP_666640.1 | 1 | 108 | ||
| GenBank mRNA | BC119106.1 | - | 1 | 198 | 8733855 | |
| BC119108.1 | - | 1 | 198 | 8733857 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr223| Gene Symbol: Olfr223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146429.1 | NP_666640.1 | 1 | 108 | ||
| GenBank mRNA | BC119106.1 | - | 1 | 198 | 8733855 | |
| BC119108.1 | - | 1 | 198 | 8733857 | ||