siRNA Details
| Gene Symbol: Olfr1428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146678.2 | NP_666889.2 | 1 | 935 | ||
| GenBank mRNA | BC119441.2 | - | 1 | 935 | 40042424 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1428| Gene Symbol: Olfr1428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146678.2 | NP_666889.2 | 1 | 935 | ||
| GenBank mRNA | BC119441.2 | - | 1 | 935 | 40042424 | |