siRNA Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007596.2 | NP_001007597.1 | 7 | 1300 | ||
| NM_001286448.1 | NP_001273377.1 | 7 | 1845 | |||
| NM_153457.7 | NP_703187.2 | 9 | 3381 | |||
| GenBank mRNA | BC027091.1 | - | 7 | 1180 | ||
| BY615773.1 | - | 1 | 144 | |||

Rtn1
Genomic Map