siRNA Details
| Gene Symbol: Tmem184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253817.1 | NP_001240746.1 | 9 | 1468 | ||
| NM_001253819.1 | NP_001240748.1 | 10 | 1510 | |||
| NM_001253820.1 | NP_001240749.1 | 7 | 1034 | |||
| NM_172608.1 | NP_766196.1 | 9 | 1514 | |||
| GenBank mRNA | AK028196.1 | - | 9 | 1472 | ||
| AK029129.1 | - | 9 | 1492 | |||
| AK048138.1 | - | 10 | 1489 | |||
| AK149366.1 | - | 9 | 1495 | |||
| AK158789.1 | - | 9 | 1515 | |||
| AK182767.1 | - | 1 | 185 | |||
| AK189937.1 | - | 1 | 185 | |||
| AK210195.1 | - | 1 | 156 | |||
| BC023899.1 | - | 1 | 139 | |||
| BC046959.1 | - | 9 | 1468 | 6314456 | ||

Tmem184b
Genomic Map