siRNA Details
| Gene Symbol: Casc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205369.1 | NP_001192298.1 | 10 | 2642 | ||
| NM_001205370.1 | NP_001192299.1 | 9 | 2474 | |||
| NM_001205371.1 | NP_001192300.1 | 10 | 2232 | |||
| NM_177054.5 | NP_796028.2 | 9 | 2543 | |||
| NM_199038.3 | NP_950239.2 | 8 | 2375 | |||
| GenBank mRNA | AK134705.1 | - | 1 | 191 | ||
| AY484583.1 | - | 9 | 2431 | |||
| AY484585.1 | - | 8 | 2237 | |||
| BC030726.1 | - | 2 | 1274 | |||
| BC034556.1 | - | 9 | 2337 | |||
| BC064446.1 | - | 10 | 2642 | 6843743 | ||

Casc4
Genomic Map