siRNA Details
| Gene Symbol: Fbxw16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177070.3 | NP_796044.2 | 9 | 1312 | ||
| GenBank mRNA | AK078661.1 | - | 8 | 1602 | ||
| BC100319.1 | - | 9 | 1296 | 30940137 | ||
| DV662133.1 | - | 4 | 589 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw16| Gene Symbol: Fbxw16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177070.3 | NP_796044.2 | 9 | 1312 | ||
| GenBank mRNA | AK078661.1 | - | 8 | 1602 | ||
| BC100319.1 | - | 9 | 1296 | 30940137 | ||
| DV662133.1 | - | 4 | 589 | |||