siRNA Details
| Gene Symbol: Fbxw16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177070.3 | NP_796044.2 | 10 | 1383 | ||
| GenBank mRNA | AK078661.1 | - | 9 | 1673 | ||
| BC100319.1 | - | 10 | 1367 | 30940137 | ||
| DV662133.1 | - | 5 | 660 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw16| Gene Symbol: Fbxw16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177070.3 | NP_796044.2 | 10 | 1383 | ||
| GenBank mRNA | AK078661.1 | - | 9 | 1673 | ||
| BC100319.1 | - | 10 | 1367 | 30940137 | ||
| DV662133.1 | - | 5 | 660 | |||