siRNA Details
| Gene Symbol: Mustn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181390.3 | NP_852055.1 | 3 | 1117 | ||
| GenBank mRNA | AK003945.1 | - | 3 | 834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mustn1| Gene Symbol: Mustn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181390.3 | NP_852055.1 | 3 | 1117 | ||
| GenBank mRNA | AK003945.1 | - | 3 | 834 | ||