siRNA Details
| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199495.2 | NP_955789.2 | 2 | 232 | ||
| XM_008767385.1 | XP_008765607.1 | 2 | 244 | |||
| GenBank mRNA | BC093375.1 | - | 2 | 191 | 7389276 | |
| CB610197.1 | - | 2 | 232 | |||
| FQ209525.1 | - | 2 | 234 | |||
| Gene Symbol: Ndufa10l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182671.2 | NP_872612.1 | 1 | 194 | ||
| GenBank mRNA | AY260163.1 | - | 1 | 194 | ||

Ndufa10
Genomic Map