siRNA Details
| Gene Symbol: Lmbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020295.3 | NP_064691.2 | 17 | 4766 | ||
| GenBank mRNA | AK004199.1 | - | 1 | 225 | ||
| BC096579.1 | - | 1 | 688 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_183120.1 | - | 225 | Chr. - - [-] on Build | ||

Lmbr1
Genomic Map