siRNA Details
| Gene Symbol: Ffar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194057.2 | NP_918946.2 | 1 | 490 | ||
| GenBank mRNA | AB095745.1 | - | 1 | 490 | ||
| BC132014.1 | - | 1 | 584 | 40130291 | ||
| BC138099.1 | - | 1 | 584 | 8861118 | ||
More Information
Other Species Targeted
Set Membership: |
|

Ffar1
Genomic Map