siRNA Details
| Gene Symbol: A330070K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198665.1 | NP_941067.1 | 1 | 192 | ||
| GenBank mRNA | AK039605.1 | - | 1 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330070K13Rik| Gene Symbol: A330070K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198665.1 | NP_941067.1 | 1 | 192 | ||
| GenBank mRNA | AK039605.1 | - | 1 | 192 | ||