siRNA Details
| Gene Symbol: BC061237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198677.1 | NP_941079.1 | 3 | 335 | ||
| GenBank mRNA | BC061237.1 | - | 3 | 335 | 6705671 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC061237| Gene Symbol: BC061237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198677.1 | NP_941079.1 | 3 | 335 | ||
| GenBank mRNA | BC061237.1 | - | 3 | 335 | 6705671 | |