siRNA Details
| Gene Symbol: SLC22A25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199352.3 | NP_955384.3 | 3 | 595 | ||
| XM_017017684.1 | XP_016873173.1 | 7 | 1764 | |||
| XM_017017685.1 | XP_016873174.1 | 8 | 1892 | |||
| XM_017017686.1 | XP_016873175.1 | 7 | 2200 | |||
| XM_017017687.1 | XP_016873176.1 | 6 | 2215 | |||
| XM_017017688.1 | XP_016873177.1 | 3 | 1153 | |||
| XM_017017689.1 | XP_016873178.1 | 4 | 1065 | |||
| XM_017017690.1 | XP_016873179.1 | 6 | 968 | |||
| XM_017017691.1 | XP_016873180.1 | 4 | 421 | |||
| XM_017017692.1 | XP_016873181.1 | 3 | 592 | |||
| XM_017017693.1 | XP_016873182.1 | 2 | 183 | |||
| XM_017017694.1 | XP_016873183.1 | 2 | 1022 | |||
| XM_017017695.1 | XP_016873184.1 | 7 | 2305 | |||
| XM_017017696.1 | XP_016873185.1 | 4 | 724 | |||
| GenBank mRNA | AK091990.1 | - | 4 | 401 | ||
| AK127610.1 | - | 3 | 470 | |||
| AY437532.1 | - | 3 | 595 | |||
| AY517500.1 | - | 4 | 401 | |||
| BC101315.1 | - | 3 | 590 | |||
| BC101316.1 | - | 3 | 595 | |||

SLC22A25
Genomic Map