siRNA Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028532.3 | NP_082808.1 | 6, 7 | 493 | ||
| NM_199455.2 | NP_955534.1 | 7, 8 | 718 | |||
| GenBank mRNA | AK006802.1 | - | 6, 7 | 480 | ||
| BC087870.1 | - | 7, 8 | 714 | 6707505 | ||
| BC104382.1 | - | 6, 7 | 492 | |||
| BC104383.1 | - | 6, 7 | 493 | |||
| BY706785.1 | - | 6, 7 | 478 | |||

Tepp
Genomic Map