siRNA Details
Gene Symbol: Tas2r124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207026.1 | NP_996909.1 | 1 | 654 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tas2r124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207026.1 | NP_996909.1 | 1 | 654 |