siRNA Details
| Gene Symbol: FAM199X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207318.3 | NP_997201.1 | 6 | 1988 | ||
| XM_005262079.2 | XP_005262136.1 | 6 | 1856 | |||
| XM_017029270.1 | XP_016884759.1 | 6 | 1557 | |||
| GenBank mRNA | AK091221.1 | - | 7 | 1835 | ||
| AK307312.1 | - | 3 | 785 | |||
| BC016683.1 | - | 4 | 1079 | |||
| BC017665.1 | - | 6 | 1843 | 3903800 | ||
| BC035886.1 | - | 5 | 1379 | |||

FAM199X
Genomic Map