siRNA Details
| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009718.1 | NP_001009718.1 | 7 | 809 | ||
| XM_008763403.1 | XP_008761625.1 | 6 | 871 | |||
| GenBank mRNA | AY651018.1 | - | 7 | 809 | ||
| Gene Symbol: Klra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763344.1 | XP_008761566.1 | 1012 | |||

Klra2
Genomic Map