siRNA Details
| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009718.1 | NP_001009718.1 | 7, 8 | 945 | ||
| GenBank mRNA | AY651018.1 | - | 7, 8 | 945 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klra2| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009718.1 | NP_001009718.1 | 7, 8 | 945 | ||
| GenBank mRNA | AY651018.1 | - | 7, 8 | 945 | ||