siRNA Details
| Gene Symbol: Kctd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009973.1 | NP_001009973.1 | 7 | 1228 | ||
| XM_006249463.2 | XP_006249525.1 | 7 | 1304 | |||
| GenBank mRNA | AY318756.2 | - | 7 | 1228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd10| Gene Symbol: Kctd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009973.1 | NP_001009973.1 | 7 | 1228 | ||
| XM_006249463.2 | XP_006249525.1 | 7 | 1304 | |||
| GenBank mRNA | AY318756.2 | - | 7 | 1228 | ||