siRNA Details
| Gene Symbol: Prss36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011560.2 | NP_001011560.2 | 7 | 875 | ||
| XM_008759913.1 | XP_008758135.1 | 7 | 1729 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss36| Gene Symbol: Prss36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011560.2 | NP_001011560.2 | 7 | 875 | ||
| XM_008759913.1 | XP_008758135.1 | 7 | 1729 | |||