siRNA Details
| Gene Symbol: Xk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012227.1 | NP_001012227.1 | 3 | 993 | ||
| XM_006256692.2 | XP_006256754.1 | 3 | 1844 | |||
| GenBank mRNA | AY534256.1 | - | 3 | 993 | ||
| BC107559.1 | - | 3 | 1100 | 7933294 | ||

Xk
Genomic Map