siRNA Details
| Gene Symbol: Xk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012227.1 | NP_001012227.1 | 3 | 1544 | ||
| GenBank mRNA | AY534256.1 | - | 3 | 1544 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xk| Gene Symbol: Xk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012227.1 | NP_001012227.1 | 3 | 1544 | ||
| GenBank mRNA | AY534256.1 | - | 3 | 1544 | ||