siRNA Details
| Gene Symbol: Xkr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012228.1 | NP_001012228.1 | 5 | 1575 | ||
| GenBank mRNA | AY534259.1 | - | 5 | 1575 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr5| Gene Symbol: Xkr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012228.1 | NP_001012228.1 | 5 | 1575 | ||
| GenBank mRNA | AY534259.1 | - | 5 | 1575 | ||