siRNA Details
| Gene Symbol: NMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012233.1 | NP_001012233.1 | 8, 9 | 446 | ||
| GenBank mRNA | AB164465.1 | - | 8, 9 | 446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NMS| Gene Symbol: NMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012233.1 | NP_001012233.1 | 8, 9 | 446 | ||
| GenBank mRNA | AB164465.1 | - | 8, 9 | 446 | ||