siRNA Details
| Gene Symbol: Plcz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012234.1 | NP_001012234.1 | 8 | 1266 | ||
| XM_006237597.2 | XP_006237659.1 | 9 | 1495 | |||
| XM_006237600.2 | XP_006237662.1 | 8 | 1371 | |||
| GenBank mRNA | AY885259.1 | - | 8 | 1266 | ||

Plcz1
Genomic Map