siRNA Details
| Gene Symbol: Rmnd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017473.1 | NP_001017473.1 | 9 | 1293 | ||
| XM_006246205.1 | XP_006246267.1 | 9 | 1321 | |||
| GenBank mRNA | BC093386.1 | - | 9 | 1293 | 7389386 | |
| Gene Symbol: LOC103689927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246315.1 | XP_006246377.1 | 9 | 1321 | ||
| XM_008767740.1 | XP_008765962.1 | 9 | 1351 | |||

Rmnd5b
Genomic Map