siRNA Details
| Gene Symbol: Gps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017477.1 | NP_001017477.1 | 9 | 1162 | ||
| XM_006246783.2 | XP_006246845.1 | 10 | 1112 | |||
| XM_006246785.1 | XP_006246847.1 | 10 | 1112 | |||
| XM_008767855.1 | XP_008766077.1 | 9 | 1187 | |||
| XM_008767856.1 | XP_008766078.1 | 9 | 1187 | |||
| GenBank mRNA | BC083699.1 | - | 9 | 1162 | 7190067 | |
| FQ233878.1 | - | 10 | 996 | |||

Gps2
Genomic Map