siRNA Details
| Gene Symbol: Tmem100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017479.1 | NP_001017479.1 | 3 | 1324 | ||
| XM_008768100.1 | XP_008766322.1 | 2 | 1962 | |||
| XM_008768101.1 | XP_008766323.1 | 3 | 1821 | |||
| XM_008768102.1 | XP_008766324.1 | 3 | 1551 | |||
| GenBank mRNA | BC092577.1 | - | 3 | 1324 | 7124708 | |
| FQ212975.1 | - | 3 | 1329 | |||

Tmem100
Genomic Map