siRNA Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017484.1 | NP_001017484.1 | 4 | 909 | ||
| XM_006248178.2 | XP_006248240.1 | 5 | 1135 | |||
| XM_006248179.2 | XP_006248241.1 | 6 | 1237 | |||
| XM_006248180.2 | XP_006248242.1 | 5 | 1184 | |||
| XM_006248181.2 | XP_006248243.1 | 5 | 1075 | |||
| XM_006248182.2 | XP_006248244.1 | 4 | 973 | |||
| GenBank mRNA | BC082082.1 | - | 4 | 909 | 7133412 | |

Lca5l
Genomic Map