siRNA Details
| Gene Symbol: Eif2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017489.1 | NP_001017489.1 | 5, 6 | 725 | ||
| XM_006249790.2 | XP_006249852.1 | 5, 6 | 749 | |||
| GenBank mRNA | BC087701.1 | - | 5, 6 | 725 | 7314085 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2d| Gene Symbol: Eif2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017489.1 | NP_001017489.1 | 5, 6 | 725 | ||
| XM_006249790.2 | XP_006249852.1 | 5, 6 | 749 | |||
| GenBank mRNA | BC087701.1 | - | 5, 6 | 725 | 7314085 | |