siRNA Details
| Gene Symbol: Tmem81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017490.1 | NP_001017490.1 | 2 | 1451 | ||
| GenBank mRNA | BC079284.1 | - | 2 | 1451 | 7134237 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem81| Gene Symbol: Tmem81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017490.1 | NP_001017490.1 | 2 | 1451 | ||
| GenBank mRNA | BC079284.1 | - | 2 | 1451 | 7134237 | |