siRNA Details
| Gene Symbol: Thegl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017498.1 | NP_001017498.1 | 8 | 1289 | ||
| XM_006250846.2 | XP_006250908.1 | 7 | 2078 | |||
| GenBank mRNA | BC087065.1 | - | 8 | 1289 | 7132384 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thegl| Gene Symbol: Thegl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017498.1 | NP_001017498.1 | 8 | 1289 | ||
| XM_006250846.2 | XP_006250908.1 | 7 | 2078 | |||
| GenBank mRNA | BC087065.1 | - | 8 | 1289 | 7132384 | |