siRNA Details
| Gene Symbol: Hormad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017501.1 | NP_001017501.1 | 3 | 375 | ||
| XM_008770340.1 | XP_008768562.1 | 3 | 470 | |||
| XM_008770341.1 | XP_008768563.1 | 5 | 627 | |||
| XM_008770342.1 | XP_008768564.1 | 4 | 544 | |||
| XM_008770343.1 | XP_008768565.1 | 3 | 326 | |||
| XM_008770344.1 | XP_008768566.1 | 3 | 467 | |||
| GenBank mRNA | BC079253.1 | - | 3 | 375 | 7133170 | |

Hormad2
Genomic Map