siRNA Details
| Gene Symbol: Hormad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017501.1 | NP_001017501.1 | 10 | 1232 | ||
| XM_008770344.1 | XP_008768566.1 | 11 | 1393 | |||
| GenBank mRNA | BC079253.1 | - | 10 | 1232 | 7133170 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hormad2| Gene Symbol: Hormad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017501.1 | NP_001017501.1 | 10 | 1232 | ||
| XM_008770344.1 | XP_008768566.1 | 11 | 1393 | |||
| GenBank mRNA | BC079253.1 | - | 10 | 1232 | 7133170 | |