siRNA Details
| Gene Symbol: Purb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017503.1 | NP_001017503.1 | 1 | 1268 | ||
| GenBank mRNA | AB182574.1 | - | 1 | 1268 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Purb| Gene Symbol: Purb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017503.1 | NP_001017503.1 | 1 | 1268 | ||
| GenBank mRNA | AB182574.1 | - | 1 | 1268 | ||