siRNA Details
| Gene Symbol: Zrsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017504.1 | NP_001017504.1 | 1 | 1351 | ||
| XM_008770435.1 | XP_008768657.1 | 2 | 3270 | |||
| XM_008770436.1 | XP_008768658.1 | 2 | 3194 | |||
| GenBank mRNA | BC086322.1 | - | 1 | 1351 | 7301896 | |

Zrsr1
Genomic Map