siRNA Details
| Gene Symbol: CXorf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184782.1 | NP_001171711.1 | 12, 13 | 2305 | ||
| NM_018015.5 | NP_060485.4 | 13, 14 | 2596 | |||
| GenBank mRNA | AK024253.1 | - | 12, 13 | 1579 | ||
| AK291730.1 | - | 13, 14 | 2545 | |||
| BC011483.1 | - | 13, 14 | 2517 | 3453700 | ||
| BC033709.1 | - | 13, 14 | 2508 | 3910777 | ||
| BC070110.1 | - | 13, 14 | 2547 | 5264812 | ||

CXorf57
Genomic Map