siRNA Details
| Gene Symbol: LOC100911664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751691.3 | XP_003751739.1 | 5 | 3451 | ||
| GenBank mRNA | FQ227695.1 | - | 1 | 135 | ||
| Gene Symbol: LOC498750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC085940.1 | - | 5 | 1736 | 7123624 | |
| BC089212.1 | - | 5 | 1534 | 7302835 | ||

LOC100911664
Genomic Map