siRNA Details
| Gene Symbol: Sertad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017513.1 | NP_001017513.1 | 2 | 1087 | ||
| XM_006228582.1 | XP_006228644.1 | 2 | 2061 | |||
| GenBank mRNA | BC091229.1 | - | 2 | 1087 | 7321753 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sertad3| Gene Symbol: Sertad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017513.1 | NP_001017513.1 | 2 | 1087 | ||
| XM_006228582.1 | XP_006228644.1 | 2 | 2061 | |||
| GenBank mRNA | BC091229.1 | - | 2 | 1087 | 7321753 | |