siRNA Details
| Gene Symbol: MGC105567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024284.1 | NP_001019455.1 | 2 | 1260 | ||
| GenBank mRNA | BC093614.1 | - | 2 | 1260 | 7309663 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC105567| Gene Symbol: MGC105567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024284.1 | NP_001019455.1 | 2 | 1260 | ||
| GenBank mRNA | BC093614.1 | - | 2 | 1260 | 7309663 | |