siRNA Details
| Gene Symbol: LOC499331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024293.1 | NP_001019464.1 | 2 | 1478 | ||
| GenBank mRNA | BC082069.1 | - | 2 | 1478 | 7132875 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499331| Gene Symbol: LOC499331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024293.1 | NP_001019464.1 | 2 | 1478 | ||
| GenBank mRNA | BC082069.1 | - | 2 | 1478 | 7132875 | |