siRNA Details
| Gene Symbol: GPATCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018025.2 | NP_060495.2 | 8 | 1201 | ||
| NR_135270.1 | 8 | 1201 | ||||
| XM_006723255.3 | XP_006723318.1 | 8 | 916 | |||
| GenBank mRNA | AF434677.1 | - | 8 | 887 | ||
| AK096005.1 | - | 8 | 895 | |||
| AK292503.1 | - | 8 | 895 | |||
| AK315712.1 | - | 8 | 913 | |||
| AL834465.1 | - | 8 | 892 | |||
| BC006108.1 | - | 8 | 1201 | 3029084 | ||

GPATCH1
Genomic Map