siRNA Details
| Gene Symbol: MGC114499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024907.1 | NP_001020078.1 | 3 | 975 | ||
| GenBank mRNA | BC097443.1 | - | 3 | 975 | 7456163 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC114499| Gene Symbol: MGC114499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024907.1 | NP_001020078.1 | 3 | 975 | ||
| GenBank mRNA | BC097443.1 | - | 3 | 975 | 7456163 | |