siRNA Details
| Gene Symbol: TSPYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003309.3 | NP_003300.1 | 1 | 2120 | ||
| GenBank mRNA | AK026849.1 | - | 1 | 981 | ||
| AL136629.1 | - | 1 | 1934 | |||
| BC048969.1 | - | 1 | 1936 | 5297724 | ||
| BC068554.1 | - | 1 | 1933 | |||

TSPYL1
Genomic Map