siRNA Details
Gene Symbol: Adam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020303.1 | NP_064699.1 | 14 | 1574 | ||
XM_006253321.2 | XP_006253383.1 | 14 | 1624 | |||
XM_006253322.2 | XP_006253384.1 | 14 | 1561 | |||
GenBank mRNA | BC079016.1 | - | 14 | 1544 | ||
BC091169.1 | - | 14 | 1574 | 7112347 | ||
Y11489.1 | - | 7 | 889 |
